Plasmid DNA mini kit, E.Z.N.A.® FastFilter
Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D6944-00 D6944-01 D6944-02
OMEGD6944-00EA 48.4 EUR
OMEGD6944-00 D6944-01 D6944-02
Plasmid DNA mini kit, E.Z.N.A.® FastFilter
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents
Isolate plasmid DNA from up to 5 ml culture in 9 minutes utilizing an innovative lysate clearance column.
  • Rapid - 9 Minute Processing time from a bacterial pellet
  • Unique - Lysate Clearance and DNA Binding in 1 step
  • Convenient - Lysate Clearance Column eliminates cellular debris pelleting and transfer step
  • Versatile - Process up to 5 ml bacterial cultures with different plasmid types and culture medias
The E.Z.N.A.® fast filter plasmid DNA mini kit is designed for rapid purification of high-quality plasmid DNA starting with bacterial pellet from up to 5 ml of culture following the alkaline-lysis method in just 9 minutes. The kit features an innovative and first of its kind fast filter mini column nested within a regular HiBind® silica mini spin column to combine lysate clearance and DNA binding into one simple centrifugation step. Following wash and elute steps, purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, transformation, PCR and DNA sequencing.