E.Z.N.A.® Plasmid Mini kit II, Q-spin (without lid)
Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D6945-01 D6945-02 D6945-00
D6945-01EA 150 EUR
D6945-01 D6945-02 D6945-00
E.Z.N.A.® Plasmid Mini kit II, Q-spin (without lid)
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents
This second generation plasmid miniprep kit yields 40 to 70 µg of high-quality DNA in less than 30 minutes using the same format as the E.Z.N.A.® Plasmid Mini Kit I. The increased DNA binding capacity of the HiBind® Mini Column II allows culture volumes of up to 15 ml to be used, thus bridging the gap between mini and midi prep protocols. Plasmid DNA can be readily and quickly isolated for most downstream applications such as routine screening, restriction enzyme digestion, and DNA sequencing.
  • Purification of plasmid DNA in 30 minutes or less
  • No phenol/chloroform extractions
  • DNA suitable for downstream applications