Demecolcine ≥98% (by HPLC), Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

D7385-10MG D7385-50MG D7385-5MG
D7385-10MGEA 413 EUR
D7385-10MG D7385-50MG D7385-5MG
Demecolcine ≥98% (by HPLC), Sigma-Aldrich®
Demecolcine

Demecolcine has been used as a microtubule depolymerizing agent. It has also been used to inhibit mitotic division.

Formule: C₂₁H₂₅NO₅
Smeltpunt: 184 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00075459
CAS nummer: 477-30-5
EINECS: 207-514-6
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3306ed62d1b783805fbc7509957d976987cbe901&vid=3306ed62d1b783805fbc7509957d976987cbe901&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (colour) Faint yellow to yellow and faint beige
Appearance (form) Powder
Purity (hplc area %) ≥98%
Solubility (colour) Faint yellow to very dark yellow and faint green-yellow to very dark green-yellow
Solubility (turbidity) Clear
Solubility (method) 10 mg plus 1 ml of chloroform
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR