Mailing boxes, EPS
Leverancier: DAKLAPACK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
520005 13005 520006 13004
DAKL520005EA 19.8 EUR
DAKL520005 DAKL13005 DAKL520006 DAKL13004
Mailing boxes, EPS
Mailer enveloppen en transportverpakkingen
Made up of expanded polystyrene (EPS).
  • Non transparent
  • Can be used for cooling elements, gel packs and perfect for dry ice
Suitable for refrigerated transport of biological substances, pharmaceuticals, and blood products.