Veiligheidsbril, Op-Tema®
Leverancier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1004949 1000016 1004947 1000017 1000018 1004948 1004960 1000020
111-1229EA 7.63 EUR
111-1229 111-3500 111-0731 DALL1000017 DALL1000018 DALL1004948 DALL1004960 DALL1000020
Veiligheidsbril, Op-Tema®
Brillen
Comfortable safety spectacles with lightweight full view lens and built-in moulded browbar.
  • Soft, adjustable nose piece
  • Fully adjustable
  • Adjustable, break-away Flexicord retainer for added safety
Weight: 28 g
Certificaten: EN 166, EN 170; 1F