Liquid detergent, concentrated, Contrad 70
Leverancier: DECON LAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1003.5L 1002
DECN1003.5LEA 225 EUR
DECN1003.5L DECN1002
Liquid detergent, concentrated, Contrad 70
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Concentrated liquid detergent which is ideal for laboratory washers and ultrasonic cleaners. For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, ferrous metal, ceramic and plastics.
  • Phosphate- and chlorine-free
  • Free-rinsing.