Laboratory glass cleaner, deconex® 22 LIQ-x
Leverancier: Borer Chemie
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
519200.00-KK5W 519200.00-K10W 519200.00-K25W
DECO519200.00-KK5WEA 89.3 EUR
DECO519200.00-KK5W DECO519200.00-K10W DECO519200.00-K25W
Laboratory glass cleaner, deconex® 22 LIQ-x
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Liquid, alkaline cleaning concentrate for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum and perfect results even at low concentrations
  • High cleaning performance due to excellent dispersing capabilities
  • Phosphate- and surfactant-free, no release of phosphates and surfactants into waste water
  • EDTA-, NTA- and chlorine-free
  • Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process
Order Now
Specificaties
Environmentally Preferable