Contact plates
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
200208 200218
DELT200208EA 237 EUR
DELT200208 DELT200218
Contact plates
Cultuurschalen Petrischalen
Manufactured from transparent PS, these contact plates are for use in the pharmaceutical industry and in hospital environments for determining bacterial contamination of surfaces such as the skin, operating tables, refrigerated gondolas, and work surfaces.
  • Aseptic production
  • Moulded grid makes it possible to identify the detected contamination per cm² and it facilitates the counting of colonies
  • Shape of the lid makes stacking perfectly stable during transport and incubation, and saves space on work surfaces
Ø×H: 65,7×14,7 mm