Jars, wide mouth, with lid and insert plug
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
202810 202802 202803 202818 202804 202839 202823.1 202828 202814 202815 202813 202820 202823 202802.1 202820.O 202828G 202813.1O 202815B.O 202802B 202814.1
215-4211EA 32.3 EUR
215-4211 215-4214 215-4215 215-4218 215-4219 DELT202839 DELT202823.1 DELT202828 DELT202814 DELT202815 DELT202813 DELT202820 DELT202823 DELT202802.1 DELT202820.O DELT202828G DELT202813.1O DELT202815B.O DELT202802B DELT202814.1
Jars, wide mouth, with lid and insert plug
Potjes Potjes voor algemeen gebruik
High density polyethylene jars, translucent, with black screw cap and insert plug.
  • Supplied uncapped, except sterile models which are supplied capped and bagged
  • Sterile versions are sterilised by irradiation
Leveringsinformatie: Upon request jars can be supplied individually wrapped. Please contact your local Avantor sales office for details.