Jars with lid
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
241010 241011 241013
DELT241010EA 3 EUR
DELT241010 DELT241011 DELT241013
Jars with lid
Potjes Potjes voor algemeen gebruik
Jars, made of PP, translucent, with lid.
  • Suitable for ailmentary use
  • Supplied uncapped
  • Good chemical and temperature resistance