Rubber washers
Leverancier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2401265 2402570 2403575 2404010
DENE2401265EA 4.6 EUR
DENE2401265 DENE2402570 DENE2403575 DENE2404010
Rubber washers
Afdichtingen Afdichtringen
These washers are made from red natural rubber.
  • Hardness 40 Shore A