Temperature and humidity loggers, Compact
Leverancier: DICKSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP125
DICKSP125EA 246 EUR
DICKSP125
Temperature and humidity loggers, Compact
Dataloggers
The data loggers is self-contained with high speed USB connectivity, high/low LED alarm, an internal sensor for recording air temperatures and a replaceable battery for accurate, easy-to-use and PC-based recording. The polycarbonate casing, compact size and rugged design allows for use in numerous applications like regulatory compliance, HVAC balancing, warehouse mapping, process control.
Certificaten: IP 20 certified protection class.
Leveringsinformatie: Supplied with CR24450 lithium battery, product comes with one year of warranty.