D-(+)-Glucose monohydrate

Leverancier: DIFCO BD
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

215530
DIFC215530EA 82 EUR
DIFC215530
D-(+)-Glucose monohydrate
D-(+)-Glucose monohydrate
Formule: C₆H₁₂O₆·H₂O
MW: 198.17 g/mol
Smeltpunt: 83
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149450
CAS nummer: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR