Glycerine

Leverancier: DIFCO BD
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

228220
DIFC228220EA 230 EUR
DIFC228220
Glycerine
Glycerine
Formule: HOCH₂CH(OH)CH₂OH
Kookpunt: 290 °C (1013 hPa)
Dichtheid: 1,26 g/cm³ (20 °C)
Vlampunt: 177 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004722
CAS nummer: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR