Sealing films, DuraSeal™
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DS2-500 DS4-250 DS4-500 DS1-150 DS5-150
DIVEDS2-500EA 99.6 EUR
DIVEDS2-500 DIVEDS4-250 DIVEDS4-500 DIVEDS1-150 DIVEDS5-150
Sealing films, DuraSeal™
Afdichtingen Afdichtfilms
This durable, heat and solvent-resistant LDPE stretch film has substantial advantages over common laboratory stretch films.
  • Durable, heat-resistant, solvent-proof
  • Chemically resistant
  • Heat resistant and microwave compatible
  • Tight sealing
  • Tear resistant
Its superior heat-resistant feature allows for containers to be incubated in boiling water baths without suffering the characteristic melting of paraffin-based alternatives. Since DuraSeal is polyethylene based and free of any adhesives, it resists essentially all caustic agents and organic solvents.
Order Now
Specificaties
Environmentally Preferable