Labels, laser Tough-Tags®
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TTLW-2016 TTSW-2240 TTGP-1050 MISL-1000 FLAG-1000 LTTM-1000 CLTT-1000 CLTT-2000 CLTT-3000 RNBW-3100 TTLW-KK-20 LTTI-1000 TTLW-2016-Y TTLW-2016-B TTLW-2016-G LTTI-2000 TTSH-1000 TTLW-2016-R RNBW-3000 TTSW-2240-R TTSW-2240-B RNBW-3200 MISL-1000-B MISL-1000-G MISL-1000-O MISL-1000-R MISL-1000-Y
817-2121EA 204 EUR
817-2121 817-2122 DIVETTGP-1050 DIVEMISL-1000 DIVEFLAG-1000 DIVELTTM-1000 DIVECLTT-1000 DIVECLTT-2000 DIVECLTT-3000 DIVERNBW-3100 DIVETTLW-KK-20 DIVELTTI-1000 DIVETTLW-2016-Y DIVETTLW-2016-B DIVETTLW-2016-G DIVELTTI-2000 DIVETTSH-1000 DIVETTLW-2016-R DIVERNBW-3000 DIVETTSW-2240-R DIVETTSW-2240-B DIVERNBW-3200 DIVEMISL-1000-B DIVEMISL-1000-G DIVEMISL-1000-O DIVEMISL-1000-R DIVEMISL-1000-Y
Labels, laser Tough-Tags®
Etiketten
These Tough-Tags® polyester labels are specially top-coated to readily accept laser printing.
  • Temperature resistant from −40 to +121 °C
  • Chemically inert, resist most organic solvents and caustic agents
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
Tough-Tags® resist boiling water baths, autoclaving and moderate freezer temperatures. Tubes labeled with Tough-Tags® easily slide in and out of centrifuge rotors without binding.