Labels for petri dishes, PetriStickers™
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PSTK-1000 PSTK-1050 PSTK-1070 PSTK-1100 PSTK-2000 PSTK-2010 PSTK-2020 PSTK-2030
DIVEPSTK-1000EA 78.9 EUR
DIVEPSTK-1000 DIVEPSTK-1050 DIVEPSTK-1070 DIVEPSTK-1100 DIVEPSTK-2000 DIVEPSTK-2010 DIVEPSTK-2020 DIVEPSTK-2030
Labels for petri dishes, PetriStickers™
Etiketten
PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably.
  • Temperature resistant from +4 to +42° C
  • Water-resistant ink
Simply remove the label's protective backing and apply it to the back of a petri dish. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.