Clax Magic Multi 70C2 Stain Remover
Leverancier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100883039
DIVY100883039EA 140 EUR
DIVY100883039
Clax Magic Multi 70C2 Stain Remover
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Clax Magic Multi is a stain remover with excellent properties to remove a variety of stains such as make-up, lipstick, ink and curry stains. The product is specially formulated for use in commercial and on premise laundries and is ready to use on most types of fabric.
  • Blend of solvents and other cleaning components
  • Consists of four innovative stain-fighting solutions