TASKI® Jontec No1 F1c Heavy-Duty Floor Stripper - No Rinse
Leverancier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
101102722
DIVY101102722EA 39.5 EUR
DIVY101102722
TASKI® Jontec No1 F1c Heavy-Duty Floor Stripper - No Rinse
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Heavy duty stripper for effective removal of floor emulsions and maintenance films from alkali resistant hard floors.
  • Highly alkaline
  • High wetting and emulsifying power
  • Fast and easy emulsion removal without rinsing
Unlike conventional strippers, its revolutionary formulation enables floors to be ready for re-coating without the need to rinse, reducing normal stripping time by up to 50%.
New Product
Order Now
Specificaties
Environmentally Preferable