Tubes and systems for active sampling
Leverancier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6733011 6733021 6400131 6728891 8101821 6400746 8101271 6728851 6728831 6728919 6400745 8101861 6400271 6400743 6400744 6400741
151-3146EA 74.1 EUR
151-3146 151-3145 DRAE6400131 DRAE6728891 DRAE8101821 DRAE6400746 DRAE8101271 DRAE6728851 DRAE6728831 DRAE6728919 DRAE6400745 DRAE8101861 DRAE6400271 DRAE6400743 DRAE6400744 DRAE6400741
Tubes and systems for active sampling
Gasbuisjes
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
  • High precision guaranteed
  • Very reliable in use
  • Improved colour depth and contrast using an optimised production method
Order Now
Specificaties
Environmentally Preferable