Pipette controller, Pipet-Aid® XL
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4-000-205 4-000-205-E
DRUM4-000-205EA 1560 EUR
DRUM4-000-205 DRUM4-000-205-E
Pipette controller, Pipet-Aid® XL
Pipetten Pipetvullers
Pipette controller for pipettes from 1 to 100 ml.
  • Longer lightweight handle lowers your arm position to reduce head, neck, and upper arm strain
  • Quiet, efficient pump reduces noise and extends operation period between charges
  • Can be charged while in use to reduce downtime
  • Three-speed settings provide more precise control for both aspirate and dispense modes
This controller for pipetting under a hood has a longer handle to reduce the amount of arm lift. The ergonomic design also includes an adjustable sliding hand rest and a removable stand whichMore Product Information