Cleanroom hoods with integrated mask, Tyvek® IsoClean®, Model IC689B WS
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
IC689BWHWS
DUPOIC689BWHWSEA 598 EUR
DUPOIC689BWHWS
Cleanroom hoods with integrated mask, Tyvek® IsoClean®, Model IC689B WS
Wegwerpkleding
These hoods with integrated mask, model IC689BWH WS, are made of Tyvek®, which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Aseptically folded
  • Ties for adjustable fit in the chest area
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (
More Product Information