1,2-Dimethoxyethaan ≥99% inhibitor-free, Reagent Grade

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

E27408-1L E27408-2.5L E27408-20L
E27408-1LEA 137 EUR
E27408-1L E27408-2.5L E27408-20L
1,2-Dimethoxyethaan ≥99% inhibitor-free, Reagent Grade
1,2-Dimethoxyethaan

  • ≥95% for general laboratory use.

Formule: C₄H₁₀O₂
Kookpunt: 85 °C (1,013 hPa)
Smeltpunt: -58 °C
Dichtheid: 0,87 g/cm³ (20 °C)
Vlampunt: -2 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008502
CAS nummer: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3,II
Merck Index: 13,03251

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Water (Karl Fischer) Max. 0.5 %
Peroxides (as H₂O₂) Max. 15 ppm
Vapor Pressure 48 mmHg ( 20 °C)
Explosion Limit 10.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR