Salt meter, SSX 210-Set
Leverancier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1340-5211
EBRO1340-5211EA 371 EUR
EBRO1340-5211
Salt meter, SSX 210-Set
Elektrochemische meters Zoutgehaltemeters
The SSX 210 salt meter is used to measure the salt content in semi-solid food products, such as meat, cold cuts, cheese, salads etc. To achieve this, the electrical conductivity is measured, since this is dependent on the salt content.
  • Measurement of the relative salt content of food products (meat, sausages, ham, cheese, salads)
  • Allows consistent taste
  • Easy operation
  • Handy and robust thanks to ABS housing
  • Automatic deactivation after 5 minutes
  • Operating temperature: +10 to +40 °C
It is important that the medium to be measured also has a waterMore Product Information