Magnetic surface temperature probes
Leverancier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1341-0640 1341-0641 1343-0640 1343-0641 1343-0644 1341-0644
EBRO1341-0640EA 458 EUR
EBRO1341-0640 EBRO1341-0641 EBRO1343-0640 EBRO1343-0641 EBRO1343-0644 EBRO1341-0644
Magnetic surface temperature probes
Temperatuursondes
These probes are used for surface temperature measurement on an extended period of ferrous containing compounds, e.g. heating plates, heating tubes, pumps, flushing tools, motors and turbines.