Organic reference standards, allopurinols, Ph. Eur.

Leverancier: EDQM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A0350000 A0350010 A0350030 A0350050 A0350020 A0350040
EDQMA0350000EA 245 EUR
EDQMA0350000 EDQMA0350010 EDQMA0350030 EDQMA0350050 EDQMA0350020 EDQMA0350040
Organic reference standards, allopurinols, Ph. Eur.
Standaarden Organische standaarden

Order Now

Specificaties

Omschrijving Allopurinol Allopurinol impurity A, (5-amino-1H-pyrazole-4-carboxamide ) Allopurinol impurity C, (5-(4H-1,2,4-triazol-4-yl)-1H-pyrazole-4-carboxamide) Allopurinol impurity E, (ethyl 5-(formylamino)-1H-pyrazole-4-carboxylate) Allopurinol impurity B, (5-formylamino-1H-pyrazole-4-carboxamide) Allopurinol impurity D, (ethyl 5-amino-1H-pyrazole-4-carboxylate)
Environmentally Preferable
CAS No. 315-30-0 27511-79-1 1346604-13-4 31055-19-3 22407-20-1 6994-25-8
Formule C₅H₄N₄O C₈H₁₄N₈O₆S C₆H₆N₆O C₇H₉N₃O₃ C₅H₆N₄O₂ C₆H₉N₃O₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR