Organic reference standard, Chlorhexidine dihydrochloride, Ph.Eur., RS

Leverancier: EDQM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C1540000
EDQMC1540000EA 245 EUR
EDQMC1540000
Organic reference standard, Chlorhexidine dihydrochloride, Ph.Eur., RS
Standaarden Organische standaarden

  • In Monograph 0659

Order Now

Specificaties

Omschrijving Chlorhexidine dihydrochloride
Environmentally Preferable
CAS No. 3697-42-5
Formule C₂₂H₃₂Cl₄N₁₀
Opslagtemperatuur Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR