L(-)-Cystine

Leverancier: EDQM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2

C3300000
EDQMC3300000EA 245 EUR
EDQMC3300000
L(-)-Cystine
L(-)-Cystine
Formule: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240.3 g/mol
Kookpunt: 428
Smeltpunt: 261…262 °C
Dichtheid: 1.677 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064228
CAS nummer: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR