Organic reference standard, DL-Methionine, Ph.Eur., RS

Leverancier: EDQM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M0950000
EDQMM0950000EA 245 EUR
EDQMM0950000
Organic reference standard, DL-Methionine, Ph.Eur., RS
Standaarden Organische standaarden

  • In Monograph 0624
  • Solubility in water: 34 g/l at 25°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR