Quinine sulphate dihydrate

Leverancier: EDQM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulphuric acid dihydrate, (-)-Quinine sulphate dihydrate

Q0200000
EDQMQ0200000EA 245 EUR
EDQMQ0200000
Quinine sulphate dihydrate
Quinine sulphate dihydrate
Formule: C₄₀H₅₀N₄O₈S·2H₂O
MW: 782.96 g/mol
Smeltpunt: 219
Dichtheid: 1.36 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150790
CAS nummer: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR