Organic reference standard, Trolamine, Ph.Eur., RS

Leverancier: EDQM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T2545000
EDQMT2545000EA 245 EUR
EDQMT2545000
Organic reference standard, Trolamine, Ph.Eur., RS
Standaarden Organische standaarden

  • In Monograph 1577
  • Solubility in water: 149 g/l at 20 °C
  • Pharmaceutical applications

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR