Organic reference standard, Amlodipine besilate, Ph.Eur., RS
Leverancier: EDQM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Y0000049
EDQMY0000049EA 245 EUR
EDQMY0000049
Organic reference standard, Amlodipine besilate, Ph.Eur., RS
Standaarden Organische standaarden
Filled under inert atmosphere protected from humidity.
  • In Monograph 1491
  • Solubility in water: 1 - 10 g/l