Experimentation kits, what is an epidemic and how does an infection spread?
Leverancier: EDVOTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S-68
EDVOS-68EA 82.5 EUR
EDVOS-68
Experimentation kits, what is an epidemic and how does an infection spread?
Educatief materiaal Educatief materiaal over biologie Physiology and Health Learning Activities
Infectious agents such as bacteria and viruses can spread rapidly through a population and cause widespready disease and death. In this experiment, students use coloured solutions to simulate the spreading of a disease in the classroom.
  • For 10 lab groups
  • Complete in 30 minutes
Leveringsinformatie: Kit includes instructions, HCl solution, NaOH, colour indicator, test tubes, and pipettes.