Dexamethasone

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C12170400
EHERC12170400EA 70.2 EUR
EHERC12170400
Dexamethasone
Dexamethasone
Formule: C₂₂H₂₉FO₅
Smeltpunt: 268…271 °C
MDL Nummer: MFCD00064136
CAS nummer: 50-02-2
EINECS: 200-003-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR