2,5-Dichloorfenol

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C12452000
EHERC12452000EA 31.7 EUR
EHERC12452000
2,5-Dichloorfenol
2,5-Dichloorfenol
Formule: C₆H₄Cl₂O
MW: 163 g/mol
Kookpunt: 211
Smeltpunt: 60…61 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002174
CAS nummer: 583-78-8
EINECS: 209-520-4
UN: 2020
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR