Organic reference standard, doxycycline hyclate, Dr. Ehrenstorfer
Leverancier: EHRENSTORFER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C13084280
EHERC13084280EA 94.8 EUR
EHERC13084280
Organic reference standard, doxycycline hyclate, Dr. Ehrenstorfer
Standaarden Organische standaarden
Certificaten: ISO/IEC 17025 certified.
Order Now
Specificaties
Omschrijving Doxycycline hyclate
Environmentally Preferable
CAS No. 24390-14-5
Formule 2C₂₂H₂₄N₂O₈·C₂H₆O·2HCl·H₂O