Fast green FCF

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C13406000
EHERC13406000EA 110 EUR
EHERC13406000
Fast green FCF
Fast green FCF
Formule: C₃₇H₃₄N₂Na₂O₁₀S₃
Smeltpunt: 290 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013053
CAS nummer: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR