Azelaic acid

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid

C14126200
EHERC14126200EA 52.9 EUR
EHERC14126200
Azelaic acid
Azelaic acid
Formule: C₉H₁₆O₄
MW: 188.22 g/mol
Kookpunt: 371
Smeltpunt: 106
Dichtheid: 1.0287 g/cm³ (20 °C)
Vlampunt: 215
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004432
CAS nummer: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR