Isobutylbenzene

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C14394600
EHERC14394600EA 48 EUR
EHERC14394600
Isobutylbenzene
Isobutylbenzene
Formule: (C₆H₅)CH₂CH(CH₃)CH₃
Kookpunt: 173 °C (1013 hPa)
Smeltpunt: –51 °C
Dichtheid: 0,8538 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008936
CAS nummer: 538-93-2
EINECS: 208-706-2
UN: 2709
ADR: 3,III
Merck Index: 13,05151

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR