Isoflucypram

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C14424700
EHERC14424700EA 0 EUR
EHERC14424700
Isoflucypram
Standaarden Organische standaarden
New Product

Order Now

Specificaties

Environmentally Preferable
CAS No. 1255734-28-1
Formule C₁₉H₂₁ClF₃N₃O
Opslagtemperatuur 20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR