Isopentylamine

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C15084500
EHERC15084500EA 38.2 EUR
EHERC15084500
Isopentylamine
Isopentylamine
Formule: C₅H₁₃N
Kookpunt: 97 °C (1013 hPa)
Smeltpunt: –68 °C
Dichtheid: 0,75 g/cm³ (20 °C)
Vlampunt: –1 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008203
CAS nummer: 107-85-7
EINECS: 203-526-0
UN: 1106
ADR: 3,II
Merck Index: 13,05128

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR