n-Pentatriacontane

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C15981700
EHERC15981700EA 110 EUR
EHERC15981700
n-Pentatriacontane
n-Pentatriacontane
Formule: H₃C(CH₂)₃₃CH₃
Kookpunt: 490 °C (1013 hPa)
Dichtheid: 0,8157 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00059397
CAS nummer: 630-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR