3,3',5,5'-Tetramethylbenzidine (TMB), certified

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

C17413000
EHERC17413000EA 51.7 EUR
EHERC17413000
3,3',5,5'-Tetramethylbenzidine (TMB), certified
3,3',5,5'-Tetramethylbenzidine (TMB)
Formule: C₁₆H₂₀N₂
Kookpunt: 402 °C (1013 hPa)
Smeltpunt: 166…170 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007748
CAS nummer: 54827-17-7
EINECS: 259-364-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR