TBPP (tris(2,3-dibromopropyl)phosphate)

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C17894340
EHERC17894340EA 71.4 EUR
EHERC17894340
TBPP (tris(2,3-dibromopropyl)phosphate)
TBPP (tris(2,3-dibromopropyl)phosphate)
Formule: C₉H₁₅Br₆O₄P
CAS nummer: 126-72-7
EINECS: 204-799-9
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR