Acenaphthene

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C20505000
EHERC20505000EA 51.7 EUR
EHERC20505000
Acenaphthene
Acenaphthene
Formule: C₁₂H₁₀
Kookpunt: 279 °C (1013 hPa)
Smeltpunt: 90…93 °C
Dichtheid: 1,024 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003807
CAS nummer: 83-32-9
EINECS: 201-469-6
UN: 3077
ADR: 9,III
Merck Index: 13,00029
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5520bcac8d97ebc91a4b9c3873c734cd4abd6427&vid=0cd04594d640995040d469a56bffa409d582f3ce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR