Acenaphthylene

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C20510000
EHERC20510000EA 71.4 EUR
EHERC20510000
Acenaphthylene
Acenaphthylene
Formule: C₁₂H₈
Kookpunt: 265 °C (1013 hPa)
Smeltpunt: 91…92 °C
Dichtheid: 0,8988 g/cm³ (20 °C)
MDL Nummer: MFCD00003806
CAS nummer: 208-96-8
EINECS: 205-917-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR