Methyl butyrate

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Butyric acid methyl ester, Butanoic acid methyl ester

CA10931780
EHERCA10931780EA 51.7 EUR
EHERCA10931780
Methyl butyrate
Methyl butyrate
Formule: C₅H₁₀O₂
Kookpunt: 102 °C (1013 hPa)
Smeltpunt: –84 °C
Dichtheid: 0,8984 g/cm³ (20 °C)
Vlampunt: 14 °C
MDL Nummer: MFCD00009391
CAS nummer: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 13,06062

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR