Organische referentiestandaard, Musk ketone 10 µg/ml in cyclohexane

Leverancier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

LA15359000CY
EHERLA15359000CYEA 62.8 EUR
EHERLA15359000CY
Organische referentiestandaard, Musk ketone 10 µg/ml in cyclohexane
Standaarden Organische standaarden

Order Now

Specificaties

Omschrijving Musk ketone
Environmentally Preferable
CAS No. 81-14-1
Formule C₁₄H₁₈N₂O₅

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR