Cole-Parmer® CN-200 Analog Heating Controllers
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
36223-02 36223-00 36223-04 36223-03
ELET36223-02EA 494 EUR
ELET36223-02 ELET36223-00 ELET36223-04 ELET36223-03
Cole-Parmer® CN-200 Analog Heating Controllers
Regelaars Temperatuurregelaars
Cost-effective control of resistive heating loads.
  • Provide manual control of uncontrolled heating mantles, tapes, and cords
  • Compact—use on benchtop or mount to a frame
Use these external one-way controllers to manually regulate the power input to your laboratory heating equipment. Each heating controller features two indicators: 'Power On' white neon light and 'Mantle/Bunsen Heater On' amber neon light. As you turn the control dial clockwise to increase power, the amber indicator light begins to pulsate. Once maximum power is reached, the light willMore Product Information