Embedding kit JB-4, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EM0100-1KT
EM0100-1KTEA 512 EUR
EM0100-1KT
Embedding kit JB-4, Sigma-Aldrich®
Kleurstoffen Microscopy and staining reagents
JB-4 is a water-soluble, GMA based, plastic resin kit intended for use in the preparation of embedded samples for high resolution light microscopy. It is a technique now widely used for research and clinical diagnosis. JB-4 yields semi-thin sections (0.5μ-2μ) with excellent morphological preservation.
  • Quality Level 200
JB-4 kit contains:
JB-4 Embedding Solution A (J4954) 800ml
JB-4 Embedding Solution B (J0330) 30 ml
Benzoyl Peroxide (J0455) 12 g
Order Now
Specificaties
Environmentally Preferable